Thermo Scientific Chemicals

Glipizide, Thermo Scientific Chemicals

Catalog number: J63398.03
1 g, Each
Thermo Scientific Chemicals

Glipizide, Thermo Scientific Chemicals

Catalog number: J63398.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63398.03
also known as J63398-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
29094-61-9
IUPAC Name
N-[2-(4-{[(cyclohexylcarbamoyl)amino]sulfonyl}phenyl)ethyl]-5-methylpyrazine-2-carboxamide
Molecular Formula
C21H27N5O4S
InChI Key
ZJJXGWJIGJFDTL-UHFFFAOYSA-N
SMILES
CC1=CN=C(C=N1)C(=O)NCCC1=CC=C(C=C1)S(=O)(=O)NC(=O)NC1CCCCC1
Assay (unspecified)
≥98.0%

Description

An ATP-dependent potassium channel blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text