Gellan Gum, Thermo Scientific Chemicals
Gellan Gum, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gellan Gum, Thermo Scientific Chemicals

An agar substitute and gelling agent
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number J63423.30
also known as J63423-30
Price (USD)/ Each
97.65
Online exclusive
108.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
97.65
Online exclusive
108.00 
Save 10.35 (10%)
Add to cart
Gellan Gum, Thermo Scientific Chemicals
Catalog numberJ63423.30
Price (USD)/ Each
97.65
Online exclusive
108.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS71010-52-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to Brown
FormPowder
Assay (special)Gel strength: ≥400 g/cm²
Assay (special)Transmittance: ≥75%
Loss on Drying≤15%
Gellan Gum is an agar substitute and gelling agent, and useful in a wide variety of immobilization matricies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Gellan Gum is an agar substitute and gelling agent. It is used in the food industry as an anti-settling agent, thickener or stabilizer. Also used in cosmetic industries due to its special gelling property. It is also used in pharmaceuticals and in tissue engineering. It is used as a substitute for agar in leptospiral media. Also useful in a wide variety of immobilization matricies.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Vieira, S.; Vial, S.; Maia, F. R.; Carvalho, M.; Reis, R. L.; Granja, P. L.; Oliveira, J. M. Gellan gum-coated gold nanorods: an intracellular nanosystem for bone tissue engineering. RSC Adv.2015, 5 (95), 77996-78005.
  2. Vora, S. R.; Bognet, B.; Patanwala, H. S.; Chinesta, F.; Ma, A. W. K. Surface Pressure and Microstructure of Carbon Nanotubes at an Air-Water Interface. Langmuir 2015, 31 (16), 4663-4672.