Thermo Scientific Chemicals

Gellan Gum, Thermo Scientific Chemicals

Catalog number: J63423.A1
1 kg, Each
Thermo Scientific Chemicals

Gellan Gum, Thermo Scientific Chemicals

Catalog number: J63423.A1
1 kg, Each
Quantity
Catalog number: J63423.A1
also known as J63423-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71010-52-1
Appearance (Color)
White to Brown
Form
Powder
Assay (special)
Gel strength: ≥400 g/cm²
Assay (special)
Transmittance: ≥75%
Loss on Drying
≤15%

Description

Gellan Gum is an agar substitute and gelling agent, and useful in a wide variety of immobilization matricies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Gellan Gum is an agar substitute and gelling agent. It is used in the food industry as an anti-settling agent, thickener or stabilizer. Also used in cosmetic industries due to its special gelling property. It is also used in pharmaceuticals and in tissue engineering. It is used as a substitute for agar in leptospiral media. Also useful in a wide variety of immobilization matricies.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text