Thermo Scientific Chemicals

Uridine-5'-triphosphate trisodium salt, 98+%

Catalog number: J63427.MC
100 mg, Each
Thermo Scientific Chemicals

Uridine-5'-triphosphate trisodium salt, 98+%

Catalog number: J63427.MC
100 mg, Each
Quantity
Catalog number: J63427.MC
also known as J63427-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19817-92-6
IUPAC Name
trisodium ({[(2R,3S,4R,5R)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)(hydrogen phosphonatooxy)phosphinate
Molecular Formula
C9H12N2Na3O15P3
InChI Key
MMJGIWFJVDOPJF-ZKGVITBDNA-K
SMILES
[Na+].[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])(=O)OP([O-])(=O)OP(O)([O-])=O)N1C=CC(=O)NC1=O
Appearance (Color)
White to cream
Form
Powder
Loss on Drying
≤15.0%
Assay (unspecified)
≥98.0%

Description

Uridine-5'-triphosphate trisodium salt, is used as an energy-rich precursor in the enzymatic biosynthesis of RNA, a potent vasodilator, and induces contractile responses in some tissues. Uridine-5?-triphosphate, Trisodium Salt is an activator of P2Y. UTP has been used to study its vasodilatory effects on the skeletal muscle cells of diabetic patients1. UTP has also been used as a purinergic agonist to study ion transport functions of P2Y2 receptors on rat basolateral membranes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Uridine-5′-triphosphate trisodium salt, is used as an energy-rich precursor in the enzymatic biosynthesis of RNA, a potent vasodilator, and induces contractile responses in some tissues. Uridine-5′-triphosphate, Trisodium Salt is an activator of P2Y. UTP has been used to study its vasodilatory effects on the skeletal muscle cells of diabetic patients1. UTP has also been used as a purinergic agonist to study ion transport functions of P2Y2 receptors on rat basolateral membranes

Solubility
Soluble in water (50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text