Thermo Scientific Chemicals

Diatrizoic acid, Thermo Scientific Chemicals

Catalog number: J63444.14
25 g, Each
Thermo Scientific Chemicals

Diatrizoic acid, Thermo Scientific Chemicals

Catalog number: J63444.14
25 g, Each
Quantity
Catalog number: J63444.14
also known as J63444-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
117-96-4
Residue on ignition
0.1% max.
Water Content (Karl Fischer Titration)
7.1% max.
Appearance (Color)
White to off-white
Form
Powder
Assay (unspecified)
98.0% min.

Description

Diatrizoic acid is a gradient reagent used for blood cell separation and diagnostic aid (radiopaque medium). Diatrizoic acid may be used as an alternative to barium sulfate for medical imaging of the gastrointestinal tract, such as upper gastrointestinal series and small bowel series.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in Methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text