Thermo Scientific Chemicals

Genistin, 99+%, Thermo Scientific Chemicals

Catalog number: J63445.MC
100 mg, Each
Thermo Scientific Chemicals

Genistin, 99+%, Thermo Scientific Chemicals

Catalog number: J63445.MC
100 mg, Each
Quantity
Catalog number: J63445.MC
also known as J63445-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
529-59-9
IUPAC Name
5-hydroxy-3-(4-hydroxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4H-chromen-4-one
Molecular Formula
C21H20O10
InChI Key
ZCOLJUOHXJRHDI-CMWLGVBASA-N
SMILES
OC[C@H]1O[C@@H](OC2=CC(O)=C3C(=O)C(=COC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@@H](O)[C@@H]1O
Form
Powder or granules
Appearance (Color)
Pale yellow
Assay from Supplier's CofA
≥99.0% (HPLC)

Description

Glucoside of genistein that inhibits protein tyrosine kinase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text