Thermo Scientific Chemicals

Finasteride, Thermo Scientific Chemicals

Catalog number: J63454.MC
100 mg, Each
Thermo Scientific Chemicals

Finasteride, Thermo Scientific Chemicals

Catalog number: J63454.MC
100 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63454.MC
also known as J63454-MC
Price (USD)
Quantity
-
Have Questions?

Description

Finasteride is used as a inhibitor of steroid 5-a-reductase, antiandrogen, an intracellular enzyme that converts the androgen testosterone into 5α dihydrotestosterone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Finasteride is used as a inhibitor of steroid 5-a-reductase, antiandrogen, an intracellular enzyme that converts the androgen testosterone into 5α dihydrotestosterone.

Solubility
Soluble in water, DMSO, ethanol, methanol, and chloroform.

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text