Thermo Scientific Chemicals
6-Cyano-7-nitro-1,4-dihydroquinoxaline-2,3-dione, 98%, Thermo Scientific Chemicals
Catalog number: J63472.MF
50 mg, Each
Thermo Scientific Chemicals
6-Cyano-7-nitro-1,4-dihydroquinoxaline-2,3-dione, 98%, Thermo Scientific Chemicals
Catalog number: J63472.MF
50 mg, Each
Quantity
Catalog number: J63472.MF
also known as J63472-MF
Price (USD)
Quantity
-
Have Questions?
Description
Potent, selective kainate/quisqualate (non-NMDA) receptor antagonist