3-Bromo-7-nitroindazole, 98+%, Thermo Scientific Chemicals
3-Bromo-7-nitroindazole, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromo-7-nitroindazole, 98+%, Thermo Scientific Chemicals

Inhibitor of nitric oxide synthase (NOS)
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number J63480.03
also known as J63480-03
Price (USD)/ Each
141.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
141.00
Add to cart
3-Bromo-7-nitroindazole, 98+%, Thermo Scientific Chemicals
Catalog numberJ63480.03
Price (USD)/ Each
141.00
-
Add to cart
Chemical Identifiers
CAS74209-34-0
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Melting Point (clear melt)183.0-190.0?C
Appearance (Color)Yellow
FormPowder
Inhibitor of nitric oxide synthase (NOS)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only