Thermo Scientific Chemicals

Sodium phosphate, 0.2M buffer soln., pH 7.0

Catalog number: J63482.AP
500 mL, Each
Thermo Scientific Chemicals

Sodium phosphate, 0.2M buffer soln., pH 7.0

Catalog number: J63482.AP
500 mL, Each
Quantity
Catalog number: J63482.AP
also known as J63482-AP
Price (USD)
Quantity
-
Have Questions?

Description

Sodium phosphate is widely used as a reagent with high buffering capacity for molecular biology, biochemistry, and chromatography

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phosphate is widely used as a reagent with high buffering capacity for molecular biology, biochemistry, and chromatography

Solubility
Fully miscible.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text