Sodium phosphate is widely used as a reagent with high buffering capacity for molecular biology, biochemistry, and chromatography
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium phosphate is widely used as a reagent with high buffering capacity for molecular biology, biochemistry, and chromatography
Solubility
Fully miscible.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
- Pamela J. Fraker.; John C. Speck Jr. Protein and cell membrane iodinations with a sparingly soluble chloroamide, 1,3,4,6-tetrachloro-3a,6a-diphenylglycoluril.Biochemical and Biophysical Research Communications.1978, (4), 849-857.
- Hochuli, E.; Dobeli, H.; Schacher, A. New metal chelate adsorbent selective for proteins and peptides containing neighbouring histidine residues.Journal of Chromatography A.1987, 177-184.