Thermo Scientific Chemicals

TBE (5X), TRIS + borate + EDTA

Catalog number: J63487.K3
2 L, Each
Thermo Scientific Chemicals

TBE (5X), TRIS + borate + EDTA

Catalog number: J63487.K3
2 L, Each
Quantity
Catalog number: J63487.K3
also known as J63487-K3
Price (USD)
Quantity
-
Have Questions?
Form
Liquid
pH
8.4 ± 0.1
Appearance (Color)
Clear colorless to pale yellow
Composition
0.45M Tris base, 0.45M Boric acid, 10mM EDTA
Comment
All components are dissolved in UV treated 18.2 megohm-cm ± 1 water, followed by pH adjustment and filtered through 1 micron filter

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text