Thermo Scientific Chemicals

Substance P, Thermo Scientific Chemicals

Catalog number: J63489.MB
25 mg, Each
Thermo Scientific Chemicals

Substance P, Thermo Scientific Chemicals

Catalog number: J63489.MB
25 mg, Each
Quantity
Catalog number: J63489.MB
also known as J63489-MB
Price (USD)
Quantity
-
Have Questions?
Appearance (Color)
White
Form
Lyophilized Powder
Assay from Supplier's CofA
Peptide Content: Report Value
Assay from Supplier's CofA
Peptide Purity: Report Value
Mass Spec
Exhibits correct MW

Description

Substance P is used as a neurokinin NK-1 receptor agonist, neurokinin undecapeptide neurotransmitter and neuromodulator, also used in cancer cell proliferation and that inhibition of this component can induce apoptosis in cancer cells and decrease the steady state of Her2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text