Chemical Identifiers
CAS12542-36-8
IUPAC Name1,1',6,6',7,7'-hexahydroxy-3,3'-dimethyl-5,5'-bis(propan-2-yl)-[2,2'-binaphthalene]-8,8'-dicarbaldehyde; acetic acid
Molecular FormulaC32H34O10
InChI KeyNIOHNDKHQHVLKA-UHFFFAOYSA-N
SMILESCC(O)=O.CC(C)C1=C(O)C(O)=C(C=O)C2=C(O)C(=C(C)C=C12)C1=C(O)C2=C(C=O)C(O)=C(O)C(C(C)C)=C2C=C1C
View more