Thermo Scientific Chemicals
Phosphate-buffered saline (PBS, 10X), with Triton∣r X-100
Catalog number: J63521.AK
250 mL, Each
Thermo Scientific Chemicals
Phosphate-buffered saline (PBS, 10X), with Triton∣r X-100
Catalog number: J63521.AK
250 mL, Each
Capacity
Catalog number: J63521.AK
also known as J63521-AK
Price (USD)
Quantity
-
Have Questions?
Description
Phosphate-buffered saline is used in biological research. PBS buffer is ideal for maintaining a constant pH, and since it is isotonic to cells. PBS is frequently used as a wash and dilution buffer, and is also used to take a reference spectrum when measuring protein adsorption. And also used in immunoassay procedures.