Flecainide Acetate, 98%, Thermo Scientific Chemicals
Flecainide Acetate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Flecainide Acetate, 98%, Thermo Scientific Chemicals

Sodium channel blocker
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63527.MC
also known as J63527-MC
Price (USD)/ Each
214.65
Online exclusive
238.00 
Save 23.35 (10%)
-
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (USD)/ Each
214.65
Online exclusive
238.00 
Save 23.35 (10%)
Add to cart
Flecainide Acetate, 98%, Thermo Scientific Chemicals
Catalog numberJ63527.MC
Price (USD)/ Each
214.65
Online exclusive
238.00 
Save 23.35 (10%)
-
Add to cart
Chemical Identifiers
CAS54143-56-5
IUPAC NameN-[(piperidin-2-yl)methyl]-2,5-bis(2,2,2-trifluoroethoxy)benzamide; acetic acid
Molecular FormulaC19H24F6N2O5
InChI KeyRKXNZRPQSOPPRN-UHFFFAOYNA-N
SMILESCC(O)=O.FC(F)(F)COC1=CC=C(OCC(F)(F)F)C(=C1)C(=O)NCC1CCCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystals or crystalline powder
Assay from Suppliers CofA≥97.5% (HPLC)
Flecainide acetate is used as a sodium channel blocker and as an anti-arrhythmic. In general, it is used to regulate heartbeats of the upper heart and lower heart chambers. It can also be used to slow down an overactive heart. It is also useful in the treatment of ventricular tachycardias and ventricular arrhythmias.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dilute acetic acid, dimethyl sulfoxide, ethanol, methanol and water.

Notes
Incompatible with oxidizing agents and heat.
RUO – Research Use Only

General References:

  1. Doki, K.; Sekiguchi, Y.; Keisuke Kuga, K.; Aonuma, K.; Homma, M.Serum flecainide S/R ratio reflects the CYP2D6 genotype and changes in CYP2D6 activity. Drug Metab. Pharmacokinet. 2015, 30 (4), 257-262.
  2. Moffett, B. S.; Valdes, S. O.; Lupo, P. J.; delaUz, C.; Miyake, C.; Krenek, M.; Kim, J. J. Flecainide Use in Children with Cardiomyopathy or Structural Heart Disease. Pediatr. Cardiol. 2015, 36 (1), 146-150.