Thermo Scientific Chemicals

Triamcinolone acetonide, 98+%, Thermo Scientific Chemicals

Catalog number: J63548.03
1 g, Each
Thermo Scientific Chemicals

Triamcinolone acetonide, 98+%, Thermo Scientific Chemicals

Catalog number: J63548.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63548.03
also known as J63548-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76-25-5
IUPAC Name
(1S,2S,4R,8S,9S,11S,12R,13S)-12-fluoro-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
Molecular Formula
C24H31FO6
InChI Key
YNDXUCZADRHECN-JNQJZLCISA-N
SMILES
CC1(C)O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
Assay (unspecified)
>98.0%

Description

Triamcinolone acetonide suppresses allergic, inflammatory, and immune responses. Intravitreal administration of triamcinolone acetonide is used against diabetic macular edema. It is also used to relieve the discomfort of mouth sores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in DMSO or ethanol. Slightly soluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, acids, heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text