Thermo Scientific Chemicals

Cetirizine dihydrochloride, 99+%, Thermo Scientific Chemicals

Catalog number: J63549.09
10 g, Each
Thermo Scientific Chemicals

Cetirizine dihydrochloride, 99+%, Thermo Scientific Chemicals

Catalog number: J63549.09
10 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63549.09
also known as J63549-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
83881-52-1
IUPAC Name
dihydrogen 2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)acetic acid dichloride
Molecular Formula
C21H27Cl3N2O3
InChI Key
PGLIUCLTXOYQMV-UHFFFAOYNA-N
SMILES
[H+].[H+].[Cl-].[Cl-].OC(=O)COCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1
Form
Powder or crystalline powder
Appearance (Color)
White to pale cream
Assay from Supplier's CofA
≥99.0% (HPLC)
pH
1.20-1.80 (5.0% solution in water)

Description

Cetirizine dihydrochloride is used as a Histamine H-1 receptor antagonist, antihistaminic, piperazines. Inhibits activation of eosinophils and neutrophils. Non-sedating type histamine H1-receptor antagonist; major metabolite of hydroxyzine. Pharmacological activity resides primarily in the (R)-isomer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cetirizine dihydrochloride is used as a Histamine H-1 receptor antagonist, antihistaminic, piperazines. Inhibits activation of eosinophils and neutrophils. Non-sedating type histamine H1-receptor antagonist; major metabolite of hydroxyzine. Pharmacological activity resides primarily in the (R)-isomer.

Solubility
Soluble in water, DMSO, ethanol, and methanol.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text