Thermo Scientific Chemicals

DL-Cystine, Thermo Scientific Chemicals

Catalog number: J63564.14
25 g, Each
Thermo Scientific Chemicals

DL-Cystine, Thermo Scientific Chemicals

Catalog number: J63564.14
25 g, Each
Quantity
Catalog number: J63564.14
also known as J63564-14
Price (USD)
Quantity
-
Have Questions?

Description

DL-cystine is used in the preparation of sulfur-containing dimeric and monomeric surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-cystine is used in the preparation of sulfur-containing dimeric and monomeric surfactants.

Solubility
Soluble in dilute acid and alkali solution, highly soluble in water, not soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text