Ethylenediamine-N,N'-diacetic acid
Ethylenediamine-N,N'-diacetic acid
Thermo Scientific Chemicals

Ethylenediamine-N,N'-diacetic acid

Acts as a chelating agent
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number J63570.14
Price (USD)/ Each
466.65
Online exclusive
518.00 
Save 51.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
466.65
Online exclusive
518.00 
Save 51.35 (10%)
Add to cart
Ethylenediamine-N,N'-diacetic acid
Catalog numberJ63570.14
Price (USD)/ Each
466.65
Online exclusive
518.00 
Save 51.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialEthylenediamine-N,N'-diacetic acid
CAS5657-17-0
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
Ethylenediamine-N,N'-diacetic acid is acts as a chelating agent, used as a haemostatic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediamine-N,N′-diacetic acid is acts as a chelating agent, used as a haemostatic.

Solubility
Soluble in warm water.

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Rong Ma.; Ramunas J. Motekaitis.; Arthur E. Martell. Stability of metal ion complexes of N,N'-bis(2-hydroxybenzyl)ethylenediamine-N,N'-diacetic acid.Inorganica Chimica Acta.1994, (1-2), 151-155.
  2. Ivan Legg, J.; Dean W. Cooke. The Stereochemistry of Some Cobalt(III) Complexes with Ethylenediamine-N,N'-diacetic Acid and Some N-Substituted Analogs.Inorg. Chem.1965, (11), 1576-1584.