Tween 20 washing buffer, 0.5% in PBS (10X)
Tween 20 washing buffer, 0.5% in PBS (10X)
Thermo Scientific Chemicals

Tween 20 washing buffer, 0.5% in PBS (10X)

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number J63596.K2
also known as J63596-K2
Price (USD)/ Each
93.65
Online exclusive
104.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
93.65
Online exclusive
104.00 
Save 10.35 (10%)
Add to cart
Tween 20 washing buffer, 0.5% in PBS (10X)
Catalog numberJ63596.K2
Price (USD)/ Each
93.65
Online exclusive
104.00 
Save 10.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Product TypeBuffer
Quantity1 L
FormLiquid
Concentration0.5% in PBS (10X)
Unit SizeEach

Tween 20 washing buffer, 0.5% in PBS (10X) is used as a wash solution for western blot membranes and microtiter plate wells in ELISA assays. It acts as a pH stabilizer in optimal formulation, salts and detergents designed to effectively remove excess material from membranes. It finds application as laboratory detergents as well as in biomedical experiments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
Ambient temperatures