Thermo Scientific Chemicals

Vinblastine Sulfate, 98%

Catalog number: J63598.MA
10 mg, Each
Thermo Scientific Chemicals

Vinblastine Sulfate, 98%

Catalog number: J63598.MA
10 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63598.MA
also known as J63598-MA
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
143-67-9
IUPAC Name
methyl (1R,9R,10S,11R,12R,19R)-11-(acetyloxy)-12-ethyl-4-[(13S,15S,17S)-17-ethyl-17-hydroxy-13-(methoxycarbonyl)-1,11-diazatetracyclo[13.3.1.0⁴,¹².0⁵,¹⁰]nonadeca-4(12),5,7,9-tetraen-13-yl]-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2(7),3,5,13-tetraene-10-carboxylate; sulfuric acid
Molecular Formula
C46H60N4O13S
InChI Key
KDQAABAKXDWYSZ-JKDPCDLQSA-N
SMILES
OS(O)(=O)=O.CC[C@]1(O)C[C@H]2CN(C1)CCC1=C(NC3=CC=CC=C13)[C@@](C2)(C(=O)OC)C1=CC2=C(C=C1OC)N(C)[C@@H]1[C@]22CCN3CC=C[C@](CC)([C@@H]23)[C@@H](OC(C)=O)[C@]1(O)C(=O)OC
Appearance (Color)
White to pale yellow or pale cream
Form
Crystals or powder or granules or crystalline powder
Assay from Supplier's CofA
≥97.5% (dry weight basis, India sourced material)
Assay from Supplier's CofA
≥97.5% (HPLC, US sourced material)
Comment
Product sourced in US or India

Description

Anticancer agent; microtubule disrupter; Induces apoptosis.Vinblastine sulfate is therapeutically used as an antineoplastic. It is also used in the treatment of Hodgkin disease, choriocarcinoma, acute and chronic leukemias.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text