Thermo Scientific Chemicals

5-Thio-D-glucose, 97+%, Thermo Scientific Chemicals

Catalog number: J63621.MB
25 mg, Each
Thermo Scientific Chemicals

5-Thio-D-glucose, 97+%, Thermo Scientific Chemicals

Catalog number: J63621.MB
25 mg, Each
Quantity
Catalog number: J63621.MB
also known as J63621-MB
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20408-97-3
IUPAC Name
(2R,3R,4S,5R)-2,3,4,6-tetrahydroxy-5-sulfanylhexanal
Molecular Formula
C6H12O5S
InChI Key
IJJLRUSZMLMXCN-SLPGGIOYSA-N
SMILES
OC[C@@H](S)[C@@H](O)[C@H](O)[C@@H](O)C=O
Thin Layer Chromatography
97.0% min.

Description

5-Thio-D-glucose is used as an inhibitor of the cellular transport of D-glucose and D-glucose mediated insulin release, it is also an Inhibitor of spermatogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Thio-D-glucose is used as an inhibitor of the cellular transport of D-glucose and D-glucose mediated insulin release, it is also an Inhibitor of spermatogenesis.

Solubility
Miscible with water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text