Thermo Scientific Chemicals

Dicumarol, Thermo Scientific Chemicals

Catalog number: J63634.14
25 g, Each
Thermo Scientific Chemicals

Dicumarol, Thermo Scientific Chemicals

Catalog number: J63634.14
25 g, Each
Quantity
Catalog number: J63634.14
also known as J63634-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
66-76-2
IUPAC Name
4-hydroxy-3-[(4-hydroxy-2-oxo-2H-chromen-3-yl)methyl]-2H-chromen-2-one
Molecular Formula
C19H12O6
InChI Key
DOBMPNYZJYQDGZ-UHFFFAOYSA-N
SMILES
OC1=C(CC2=C(O)C3=CC=CC=C3OC2=O)C(=O)OC2=CC=CC=C12
Melting Point
289 to 291?C
Appearance (Color)
White to off white
Identification (FTIR)
Conforms to structure
Form
Powder
Loss on Drying
≤ 0.5%

Description

Dicumarol is a natural chemical used as an anticoagulant agents that functions as a vitamin K antagonist and is also a derivative of Coumarin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicumarol is a natural chemical used as an anticoagulant agents that functions as a vitamin K antagonist and is also a derivative of Coumarin.

Solubility
Soluble in aqueous alkaline solutions, organic bases, 0.1 N NaOH (15 mg/ml), Pyridine (50 mg/ml), chloroform (slightly soluble), and benzene (slightly soluble). Insoluble in water, and alcohols.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text