Thermo Scientific Chemicals

Chlorpromazine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: J63659.22
100 g, Each
Thermo Scientific Chemicals

Chlorpromazine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: J63659.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63659.22
also known as J63659-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69-09-0
IUPAC Name
hydrogen [3-(2-chloro-10H-phenothiazin-10-yl)propyl]dimethylamine chloride
Molecular Formula
C17H20Cl2N2S
InChI Key
FBSMERQALIEGJT-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(Cl)C=C12
Form
Crystals or powder or crystalline powder
Mass Spec
Consistent with structure
Appearance (Color)
White to pale cream
Loss on Drying
≤0.5%
Assay from Supplier's CofA
≥98.0% (HPLC)

Description

Chlorpromazine hydrochloride is used as a substitute for benzidine, o-dianisidine and o-tolidine in the determination of microquantities of hemoglobin and peroxidase. Chlorpromazine, Hydrochloride is used in cell signaling and neuroscience research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorpromazine hydrochloride is used as a substitute for benzidine, o-dianisidine and o-tolidine in the determination of microquantities of hemoglobin and peroxidase. Chlorpromazine, Hydrochloride is used in cell signaling and neuroscience research.

Solubility
Soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text