Thermo Scientific Chemicals

Bexarotene, 98%, Thermo Scientific Chemicals

Catalog number: J63701.EXH
300 mg, Each
Thermo Scientific Chemicals

Bexarotene, 98%, Thermo Scientific Chemicals

Catalog number: J63701.EXH
300 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63701.EXH
also known as J63701-!H
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
153559-49-0
Form
Powder or crystals or crystalline powder
Proton NMR
Conforms
Assay from Suppliers CofA
≥97.5%
Appearance (Color)
White to pale yellow

Description

Bexarotene is used as a Synthetic retinoid analog with specific affinity for the retinoid X receptor, an antineoplastic agent, already approved as an oral antineoplastic agent for cutaneous T cell lymphoma and being investigated against other cancers. A study has found that bexarotene in a mouse Alzheimer's model lowered the most toxic form of β-amyloid peptide and increased cognitive ability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bexarotene is used as a Synthetic retinoid analog with specific affinity for the retinoid X receptor, an antineoplastic agent, already approved as an oral antineoplastic agent for cutaneous T cell lymphoma and being investigated against other cancers. A study has found that bexarotene in a mouse Alzheimer′s model lowered the most toxic form of β-amyloid peptide and increased cognitive ability.

Solubility
Soluble in DMSO and ethanol

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text