Thermo Scientific Chemicals

L-Glutathione, oxidized, Thermo Scientific Chemicals

Catalog number: J63715.03
1 g, Each
Thermo Scientific Chemicals

L-Glutathione, oxidized, Thermo Scientific Chemicals

Catalog number: J63715.03
1 g, Each
Quantity
Catalog number: J63715.03
also known as J63715-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27025-41-8
IUPAC Name
(2S)-2-amino-4-{[(1R)-2-{[(2R)-2-[(4S)-4-amino-4-carboxybutanamido]-2-[(carboxymethyl)carbamoyl]ethyl]disulfanyl}-1-[(carboxymethyl)carbamoyl]ethyl]carbamoyl}butanoic acid
Molecular Formula
C20H32N6O12S2
InChI Key
YPZRWBKMTBYPTK-IOKZUGQQNA-N
SMILES
N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O
Assay (HPLC)
≥98.0%
Form
Powder
Water Content (Karl Fischer Titration)
≤8.0%
Appearance (Color)
White to pale cream

Description

L-Glutathione is used as a Acts as hydrogen acceptor in enzymatic determination of NADP and NADPH, it is used to reduced back to glutathione (GSH) via the NADPH-dependent enzyme glutathione reductase. The ratio of GSH to GSSG is often used to gauge the level of oxidative stress in cells, with higher concentrations of GSSG implying more oxidative stress

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutathione is used as a Acts as hydrogen acceptor in enzymatic determination of NADP and NADPH, it is used to reduced back to glutathione (GSH) via the NADPH-dependent enzyme glutathione reductase. The ratio of GSH to GSSG is often used to gauge the level of oxidative stress in cells, with higher concentrations of GSSG implying more oxidative stress

Solubility
Soluble in water, DMF, and ethanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text