Thermo Scientific Chemicals

N,N,N',N'-Tetramethylethylenediamine, Electrophoresis Grade

Catalog number: J63734.AE
100 mL, Each
Thermo Scientific Chemicals

N,N,N',N'-Tetramethylethylenediamine, Electrophoresis Grade

Catalog number: J63734.AE
100 mL, Each
Quantity
Catalog number: J63734.AE
also known as J63734-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-18-9
IUPAC Name
[2-(dimethylamino)ethyl]dimethylamine
Molecular Formula
C6H16N2
InChI Key
KWYHDKDOAIKMQN-UHFFFAOYSA-N
SMILES
CN(C)CCN(C)C
Appearance (Color)
Colorless
Assay (unspecified)
99.0% min.
Form
Liquid

Description

N,N,N',N'-Tetramethylethylenediamine is used as polymerization accelerator in gel electrophoresis, solvent and oxidizing reagent. N,N,N',N'-Tetramethylethylenediamine is used as anti-hyperlipidemic, plant growth stimulator, pathogen growth inhibitor, confers plant disease resistance. TEMED is used with ammonium persulfate to catalyze the polymerization of acrylamide when making polyacrylamide gels, used in gel electrophoresis. TMEDA is widely employed as a ligand for metal ions. TEMED can also be a component of Hypergolic propellants. Tetramethylethylenediamine (TEMED) is an essential catalyst for polyacrylamide gel polymerization. TEMED is frequently used with another catalyst, APS, for the preparation of polyacrylamide gels for protein and nucleic acid analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N,N′,N′-Tetramethylethylenediamine is used as polymerization accelerator in gel electrophoresis, solvent and oxidizing reagent. N,N,N′,N′-Tetramethylethylenediamine is used as anti-hyperlipidemic, plant growth stimulator, pathogen growth inhibitor, confers plant disease resistance. TEMED is used with ammonium persulfate to catalyze the polymerization of acrylamide when making polyacrylamide gels, used in gel electrophoresis. TMEDA is widely employed as a ligand for metal ions. TEMED can also be a component of Hypergolic propellants. Tetramethylethylenediamine (TEMED) is an essential catalyst for polyacrylamide gel polymerization. TEMED is frequently used with another catalyst, APS, for the preparation of polyacrylamide gels for protein and nucleic acid analysis.

Solubility
Soluble in water 10 mg/mL at 20°C.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides, copper, mercury, water, moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text