TRIS, 0.5M buffer soln., pH 6.8
TRIS, 0.5M buffer soln., pH 6.8
TRIS, 0.5M buffer soln., pH 6.8
TRIS, 0.5M buffer soln., pH 6.8
Thermo Scientific Chemicals

TRIS, 0.5M buffer soln., pH 6.8

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number J63735.K2
also known as J63735-K2
Price (USD)
160.00
Each
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)
160.00
Each
Add to cart
Specifications
CAS77-86-1
ChEBICHEBI:9754
Chemical Name or MaterialTRIS buffer soln.
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol
InChI KeyLENZDBCJOHFCAS-UHFFFAOYSA-N
View more
Ask our AI about this Product
TRIS is used as a standard biological buffer in less critical applications where economy is an important consideration, component of extraction buffer e.g. phenol extraction of DNA or RNA; and as a component of separating and stacking gels in the characterization of in vitro translation products by SDS-PAGE.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TRIS is used as a standard biological buffer in less critical applications where economy is an important consideration, component of extraction buffer e.g. phenol extraction of DNA or RNA; and as a component of separating and stacking gels in the characterization of in vitro translation products by SDS-PAGE.

Solubility
Difficult to mix.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link