Thermo Scientific Chemicals

Sodium hydroxide, 10N aq. soln., Thermo Scientific Chemicals

Catalog number: J63736.AE
100 mL, Each
Thermo Scientific Chemicals

Sodium hydroxide, 10N aq. soln., Thermo Scientific Chemicals

Catalog number: J63736.AE
100 mL, Each
Quantity
Catalog number: J63736.AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1310-73-2
IUPAC Name
sodium hydroxide
Molecular Formula
HNaO
InChI Key
HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES
[OH-].[Na+]
Appearance (Color)
Clear
Form
Liquid
Concentration
NaOH: 10M (10N)

Description

Sodium hydroxide is used to neutralize acids and make sodium salts. Sodium hydroxide is used to manufacture soaps, rayon, paper, explosives, dyestuffs, and petroleum products. It is also used in processing cotton fabric, laundering and bleaching, metal cleaning and processing, oxide coating, electroplating, and electrolytic extracting. It is commonly present in commercial drain and oven cleaners.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text