Thermo Scientific Chemicals

Potassium chloride, 1M aq. soln., Thermo Scientific Chemicals

Catalog number: J63739.AE
100 mL, Each
Thermo Scientific Chemicals

Potassium chloride, 1M aq. soln., Thermo Scientific Chemicals

Catalog number: J63739.AE
100 mL, Each
Quantity
Catalog number: J63739.AE
also known as J63739-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7447-40-7
IUPAC Name
potassium chloride
Molecular Formula
ClK
InChI Key
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES
[Cl-].[K+]
Form
Liquid
Appearance (Color)
Clear colorless
Assay from Suppliers CofA
0.95-1.05M

Description

Potassium chloride 1M solution is used as conductive standard and as electrolyte in analytical chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text