4',7-Dihydroxyisoflavone, 98+%, Thermo Scientific Chemicals
4',7-Dihydroxyisoflavone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4',7-Dihydroxyisoflavone, 98+%, Thermo Scientific Chemicals

Analog of genistein and an agonist at estrogen recpetors
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
100 mg
500 mg
Catalog number J63763.03
also known as J63763-03
Price (USD)/ Each
183.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
183.00
Add to cart
4',7-Dihydroxyisoflavone, 98+%, Thermo Scientific Chemicals
Catalog numberJ63763.03
Price (USD)/ Each
183.00
-
Add to cart
Chemical Identifiers
CAS486-66-8
IUPAC Name7-hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
Molecular FormulaC15H10O4
InChI KeyZQSIJRDFPHDXIC-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C1=COC2=CC(O)=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline solid
Identification (FTIR)Conforms to structure
Assay (HPLC)≥ 98.0%
lambda-max (UV-Vis)249, 301 nm
Analog of genistein and an agonist at estrogen recpetors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only