Thermo Scientific Chemicals

4',7-Dihydroxyisoflavone, 98+%, Thermo Scientific Chemicals

Catalog number: J63763.MC
100 mg, Each
Thermo Scientific Chemicals

4',7-Dihydroxyisoflavone, 98+%, Thermo Scientific Chemicals

Catalog number: J63763.MC
100 mg, Each
Quantity
Catalog number: J63763.MC
also known as J63763-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
486-66-8
IUPAC Name
7-hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
Molecular Formula
C15H10O4
InChI Key
ZQSIJRDFPHDXIC-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=C1)C1=COC2=CC(O)=CC=C2C1=O
Form
Crystalline solid
Identification (FTIR)
Conforms to structure
Assay (HPLC)
≥ 98.0%
lambda-max (UV-Vis)
249, 301 nm

Description

Analog of genistein and an agonist at estrogen recpetors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text