Sotalol hydrochloride, 98%
Sotalol hydrochloride, 98%
Sotalol hydrochloride, 98%
Sotalol hydrochloride, 98%
Thermo Scientific Chemicals

Sotalol hydrochloride, 98%

Potent beta adrenergic antagonist | CAS: 959-24-0 | C12H21ClN2O3S | 308.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63772.MC
also known as J63772-MC
Price (USD)
60.65
Special offer
Online exclusive
Ends: 26-Sep-2025
74.80
Save 14.15 (19%)
Each
-
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (USD)
60.65
Special offer
Online exclusive
Ends: 26-Sep-2025
74.80
Save 14.15 (19%)
Each
Add to cart
Chemical Identifiers
CAS959-24-0
SpecificationsSpecification SheetSpecification Sheet
FormWhite to pale yellow
Loss on Drying≤0.5%
Assay (unspecified)99.0 - 101.0% (Calculated on dry basis)
Ask our AI about this Product
Sotalol hydrochloride is used as a potent beta adrenergic antagonist, prolongs the action potential and increases the refractory period. Sotalol hydrochloride is also considered a non-selective β blocker and a potassium channel blocker with an IC50 of 43 μM.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sotalol hydrochloride is used as a potent beta adrenergic antagonist, prolongs the action potential and increases the refractory period. Sotalol hydrochloride is also considered a non-selective β blocker and a potassium channel blocker with an IC50 of 43 µM.

Solubility
Soluble in phosphate buffered saline, DMSO, ethanol, water, and methanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Ward, D.E.; Camm, A.J.; Spurrell, A.J. The acute cardiac electrophysiological effects of intravenous sotalol hydrochloride. Clinical Cardiology. 1979, 2, (3), 185-191.
  2. Axel Alt.; Henrike Potthast.; Juergen Moessinger.; Barbara Sickmüller.; Helga Oeser. Biopharmaceutical characterization of sotalol-containing oral immediate release drug products. Clinical Cardiology. 2004, 58, (1), 145-150.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link