Thermo Scientific Chemicals

Diacerein, Thermo Scientific Chemicals

Catalog number: J63775.03
1 g, Each
Thermo Scientific Chemicals

Diacerein, Thermo Scientific Chemicals

Catalog number: J63775.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63775.03
also known as J63775-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13739-02-1
IUPAC Name
4,5-bis(acetyloxy)-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid
Molecular Formula
C19H12O8
InChI Key
TYNLGDBUJLVSMA-UHFFFAOYSA-N
SMILES
CC(=O)OC1=CC=CC2=C1C(=O)C1=C(OC(C)=O)C=C(C=C1C2=O)C(O)=O
Assay from Supplier's CofA
≥95.0% (HPLC)
Appearance (Color)
Yellow
Form
Powder

Description

Interleukin-1 inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text