Diethylaminoethyl dextran, Thermo Scientific Chemicals
Diethylaminoethyl dextran, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethylaminoethyl dextran, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number J63781.22
also known as J63781-22
Price (USD)/ Each
176.65
Online exclusive
196.00 
Save 19.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
176.65
Online exclusive
196.00 
Save 19.35 (10%)
Add to cart
Diethylaminoethyl dextran, Thermo Scientific Chemicals
Catalog numberJ63781.22
Price (USD)/ Each
176.65
Online exclusive
196.00 
Save 19.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9015-73-0
IUPAC NameDiethylaminoethyl dextran
MDL NumberMFCD00081561
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Loss on Drying<10.0%
Total Nitrogen3.0 to 3.8%
pH(aq soln.): 5.0 to 7.0
Viscosity0.4 to 0.6
View more
DEAE-Dextran is used in the development of sustained release protein delivery and nucleic acid transfection systems. It has been speculated that DEAE-Dextran may cause a stimulation of the helper T-cell function in antibody synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DEAE-Dextran is used in the development of sustained release protein delivery and nucleic acid transfection systems. The use of DEAE-Dextran as an efficient adjuvant in vaccine production is well established. For instance, DEAE-Dextran has been used for immunization of guinea pigs and swine with inactivated foot-and-mouth disease and rhesus monkeys with inactivated Venezulean equine encephalomyelitis virus, IVEE. The mechanism of action of DEAE-Dextran is not clear. The effect on the humoral response of rhesus monkeys to IVEE vaccine results in a typical IgM-IgG response. It has been speculated that DEAE-Dextran may cause a stimulation of the helper T-cell function in antibody synthesis.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. James H. McCutchan,; Joseph S. Pagano. Enhancement of the Infectivity of Simian Virus 40 Deoxyribonucleic Acid With Diethylaminoethyl-Dextran2. J Natl Cancer Inst. 1968, 41 (2),351-357..
  2. H Duc-Nguyen. Enhancing effect of diethylaminoethyl-dextran on the focus-forming titer of a murine sarcoma virus (Harvey strain). J Virol. 1968, 2 (6), 643-644.