Thermo Scientific Chemicals

Methyl caffeate, Thermo Scientific Chemicals

Catalog number: J63786.MD
250 mg, Each
Thermo Scientific Chemicals

Methyl caffeate, Thermo Scientific Chemicals

Catalog number: J63786.MD
250 mg, Each
Quantity
Catalog number: J63786.MD
also known as J63786-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3843-74-1
IUPAC Name
methyl (2E)-3-(3,4-dihydroxyphenyl)prop-2-enoate
Molecular Formula
C10H10O4
InChI Key
OCNYGKNIVPVPPX-HWKANZROSA-N
SMILES
COC(=O)C=CC1=CC=C(O)C(O)=C1
Form
Powder
Appearance (Color)
Off-white to pale yellow
Assay (HPLC)
>/= 97.5%

Description

Inhibitor of protein tyrosine kinase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text