Description
Triethanolamine (TEA), an emulsifier, surfactant and capping agent, may be used to study its toxicity profile and mechanisms. TEA may be used in various drug delivery constructs to improve drug solubility. TEA may be used to cap CdTe nanoparticles.Triethanolamine is found are liquid laundry detergents, dishwashing liquids, general cleaners, hand cleaners, polishes, metalworking fluids, paints, shaving cream and printing inks. Triethanolamine is also used as organic additive in the grinding of cement clinker. TEA has also been used in photographic (silver halide) processing. It has been promoted as a useful alkali by amateur photographers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Triethanolamine (TEA), an emulsifier, surfactant and capping agent, may be used to study its toxicity profile and mechanisms. TEA may be used in various drug delivery constructs to improve drug solubility. TEA may be used to cap CdTe nanoparticles.Triethanolamine is found are liquid laundry detergents, dishwashing liquids, general cleaners, hand cleaners, polishes, metalworking fluids, paints, shaving cream and printing inks. Triethanolamine is also used as organic additive in the grinding of cement clinker. TEA has also been used in photographic (silver halide) processing. It has been promoted as a useful alkali by amateur photographers.
Solubility
Soluble in water.1 M, clear, colorless
Notes
Air Sensitive & Hygroscopic.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from water, moisture, air, acids, metals, halocarbons.
RUO – Research Use Only