Hexokinase, yeast
Thermo Scientific Chemicals
Hexokinase, yeast
Used to catalyzes the phosphorylation of D-hexose sugars at the C6 position utilizing, ATP as a phosphate source. Used to determine glucose, fructose, mannose and ATP.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kU
10 kU
Catalog number J63795.4I
also known as J63795-4I
Price (USD)/ Each
140.00
-
Add to cart
Quantity:
1 kU
Request bulk or custom format
Price (USD)/ Each
140.00
Add to cart
Hexokinase, yeast
Catalog numberJ63795.4I
Price (USD)/ Each
140.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Recommended StorageStore at -20°C
EINECS Number232-611-5
SensitivityHygroscopic
Quantity1 kU
FormLyophilized
View more
Hexokinase is used to catalyzes the phosphorylation of D-hexose sugars at the C6 position utilizing, ATP as a phosphate source. Used to determine glucose, fructose, mannose and ATP.