2-Chloroadenosine, Thermo Scientific Chemicals
2-Chloroadenosine, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloroadenosine, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number J63810.MD
also known as J63810-MD
Price (USD)/ Each
162.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
162.00
Add to cart
2-Chloroadenosine, Thermo Scientific Chemicals
Catalog numberJ63810.MD
Price (USD)/ Each
162.00
-
Add to cart
Chemical Identifiers
CAS146-77-0
IUPAC Name(2R,3R,4S,5R)-2-(6-amino-2-chloro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular FormulaC10H12ClN5O4
InChI KeyBIXYYZIIJIXVFW-SBTUYZBDNA-N
SMILESNC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC(Cl)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
Assay from Supplier's CofA≥98.5% (HPLC)
2-Chloroadenosine is used as a selective A1-adenosine receptor agonist, and also Induces apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroadenosine is used as a selective A1-adenosine receptor agonist, and also Induces apoptosis.

Solubility
Soluble in water.

Notes
protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Evans, M.C.; Swan, J.H.; Meldrum, B.S. An adenosine analogue, 2-chloroadenosine, protects against long term development of ischaemic cell loss in the rat hippocampus. Neuroscience Letters. 1987, 83, (3), 287-292.
  2. Haslam, R.J.; Lynham, J.A. Activation and inhibition of blood platelet adenylate cyclase by adenosine or by 2-chloroadenosine. Life Sciences. 1972, 11, (23), 1143-1154.