Thermo Scientific Chemicals

Harmol hydrochloride dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: J63827.03
1 g, Each
Thermo Scientific Chemicals

Harmol hydrochloride dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: J63827.03
1 g, Each
Quantity
Catalog number: J63827.03
also known as J63827-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
149022-16-2
IUPAC Name
hydrogen 1-methyl-2H,7H,9H-pyrido[3,4-b]indol-7-one chloride
Molecular Formula
C12H11ClN2O
InChI Key
RBOUBJPHXSVUTH-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CC1=C2NC3=CC(=O)C=CC3=C2C=CN1
Assay (unspecified)
>97.5%
Form
Powder or crystalline substance
Comment
Source is Peganum harmla seeds

Description

Harmol hydrochloride dihydrate is used to induces apoptosis by Caspase-8 activation, CNS stimulant, may act through NMDA receptors, and antitumor agent. Harmol has been shown to inhibit haspin kinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Harmol hydrochloride dihydrate is used to induces apoptosis by Caspase-8 activation, CNS stimulant, may act through NMDA receptors, and antitumor agent. Harmol has been shown to inhibit haspin kinase.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text