Thermo Scientific Chemicals

Alamethicin, Thermo Scientific Chemicals

Catalog number: J63829.LB0
5 mg, Each
Thermo Scientific Chemicals

Alamethicin, Thermo Scientific Chemicals

Catalog number: J63829.LB0
5 mg, Each
Quantity
Catalog number: J63829.LB0
also known as J63829-#0
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
59588-86-2
IUPAC Name
(4S)-4-{[(1S)-3-carbamoyl-1-{[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamoyl}propyl]carbamoyl}-4-(2-{2-[(2S)-2-{[(2S)-1-{2-[(2S)-2-(2-{2-[(2S)-2-{2-[(2S)-4-carbamoyl-2-[(2S)-2-{2-[(2S)-2-(2-{[(2S)-1-(2-acetamido-2-methylpropanoyl)pyrrolidin-2-yl]formamido}-2-methylpropanamido)propanamido]-2-methylpropanamido}propanamido]butanamido]-2-methylpropanamido}-3-methylbutanamido]-2-methylpropanamido}acetamido)-4-methylpentanamido]-2-methylpropanoyl}pyrrolidin-2-yl]formamido}-3-methylbutanamido]-2-methylpropanamido}-2-methylpropanamido)butanoic acid
Molecular Formula
C92H150N22O25
InChI Key
LGHSQOCGTJHDIL-UTXLBGCNSA-N
SMILES
CC(C)C[C@H](NC(=O)CNC(=O)C(C)(C)NC(=O)[C@@H](NC(=O)C(C)(C)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](C)NC(=O)C(C)(C)NC(=O)[C@H](C)NC(=O)C(C)(C)NC(=O)[C@@H]1CCCN1C(=O)C(C)(C)NC(C)=O)C(C)C)C(=O)NC(C)(C)C(=O)N1CCC[C@H]1C(=O)N[C@@H](C(C)C)C(=O)NC(C)(C)C(=O)NC(C)(C)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@H](CO)CC1=CC=CC=C1
Form
Powder
Solution Test
DMSO, 10mg/ml: clear colorless
Appearance (Color)
White to pale cream
Assay from Supplier's CofA
≥95.0%
Solution Test
Methanol, 10mg/ml: clear colorless

Description

Alamethicin is used as a channel-forming ionophore that activates membrane enzymes, membrane channel-forming peptide. It is a 20-amino acid channel-forming peptide antibiotic that can function as a monovalent cation ionophore. Alamethicin has been used to develop methods for the routine assessment of potential new drug candidates to elicit their pharmacokinetic drug interactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alamethicin is used as a channel-forming ionophore that activates membrane enzymes, membrane channel-forming peptide. It is a 20-amino acid channel-forming peptide antibiotic that can function as a monovalent cation ionophore. Alamethicin has been used to develop methods for the routine assessment of potential new drug candidates to elicit their pharmacokinetic drug interactions.

Solubility
Soluble in DMSO and ethanol.

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text