Methoxychlor, Thermo Scientific Chemicals
Methoxychlor, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methoxychlor, Thermo Scientific Chemicals

Molecule with estrogenic activity
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number J63839.14
also known as J63839-14
Price (USD)/ Each
211.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
211.00
Add to cart
Methoxychlor, Thermo Scientific Chemicals
Catalog numberJ63839.14
Price (USD)/ Each
211.00
-
Add to cart
Chemical Identifiers
CAS72-43-5
IUPAC Name1-methoxy-4-[2,2,2-trichloro-1-(4-methoxyphenyl)ethyl]benzene
Molecular FormulaC16H15Cl3O2
InChI KeyIAKOZHOLGAGEJT-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C(C1=CC=C(OC)C=C1)C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Transition Temperature84.0-90.0°C
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥94.0%
Molecule with estrogenic activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only