Dihydroergotamine methanesulfonate, Thermo Scientific Chemicals
Dihydroergotamine methanesulfonate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dihydroergotamine methanesulfonate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
100 mg
Catalog number J63840.03
also known as J63840-03
Price (USD)/ Each
509.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
509.00
Add to cart
Dihydroergotamine methanesulfonate, Thermo Scientific Chemicals
Catalog numberJ63840.03
Price (USD)/ Each
509.00
-
Add to cart
Chemical Identifiers
CAS6190-39-2
IUPAC Name(2R,4R,7R)-N-[(1S,2S,4R,7S)-7-benzyl-2-hydroxy-4-methyl-5,8-dioxo-3-oxa-6,9-diazatricyclo[7.3.0.0²,⁶]dodecan-4-yl]-6-methyl-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),9,12,14-tetraene-4-carboxamide; methanesulfonic acid
Molecular FormulaC34H41N5O8S
InChI KeyADYPXRFPBQGGAH-UMYZUSPBSA-N
SMILESCS(O)(=O)=O.CN1C[C@@H](C[C@H]2[C@H]1CC1=CNC3=CC=CC2=C13)C(=O)N[C@]1(C)O[C@@]2(O)[C@@H]3CCCN3C(=O)[C@H](CC3=CC=CC=C3)N2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥96.0% (HPLC)
Optical Rotation-17.0 to -23.0°
Appearance (Color)White to pale yellow to pale red
Loss on Drying≤2.0%
Agonist at vascular serotonin receptors; partial agonist at alpha-adrenergic and dopamine D2 receptors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Agonist at vascular serotonin receptors
  • Partial agonist at alpha-adrenergic and dopamine D2 receptors
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only