Agarose gel loading dye (6X), alkaline is used in DNA gel analysis in alkaline condition. It is used in agarose gel electrophoresis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Agarose gel loading dye (6X), alkaline is used in DNA gel analysis in alkaline condition. It is used in agarose gel electrophoresis.
Solubility
Fully miscible in water.
Notes
0.25% bromophenol blue, 0.25% xylene cyanol FF and 40% glycerol
RUO – Research Use Only
Georges F. Carle and Maynard V. Olson. Separation of chromosomal DNA molecules from yeast by orthogonal-field-alternation gel electrophoresis. Nucleic Acids Research.1984, 12 (14), 5647-5664.
Robert P. Noble. Electrophoretic separation of plasma lipoproteins in agarose gel. The Journal of Lipid Research.1968, 9 693-700.