Thermo Scientific Chemicals

2'-Deoxyadenosine, 99%

Catalog number: J63886.06
5 g, Each
Thermo Scientific Chemicals

2'-Deoxyadenosine, 99%

Catalog number: J63886.06
5 g, Each
Quantity
Catalog number: J63886.06
also known as J63886-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
958-09-8
IUPAC Name
(2R,3S,5R)-5-(6-amino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Molecular Formula
C10H13N5O3
InChI Key
OLXZPDWKRNYJJZ-RRKCRQDMSA-N
SMILES
NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1
Water Content (Karl Fischer Titration)
8.0% max
Appearance (Color)
White to off-white
Assay (HPLC)
98.5% min
Form
Powder

Description

2'-Deoxyadenosine is used in paradoxical stimulation study of human sperm motility. It plays an essential role as sperm motility stimulants used in sperm micro-injection on mouse and human embryo development. It finds an application in some cells as an energy source under energy stress conditions. It is involved in various biological processes to compare the functions of adenosine analogues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxyadenosine is used in paradoxical stimulation study of human sperm motility. It plays an essential role as sperm motility stimulants used in sperm micro-injection on mouse and human embryo development. It finds an application in some cells as an energy source under energy stress conditions. It is involved in various biological processes to compare the functions of adenosine analogues.

Solubility
Soluble in water and caustic soda.

Notes
Air sensitive. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text