Clotrimazole, Thermo Scientific Chemicals
Clotrimazole, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Clotrimazole, Thermo Scientific Chemicals

Specific inhibitor of calcium-activated potassium channels
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63895.06
also known as J63895-06
Price (USD)/ Each
77.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
77.90
Add to cart
Clotrimazole, Thermo Scientific Chemicals
Catalog numberJ63895.06
Price (USD)/ Each
77.90
-
Add to cart
Specifications
Chemical Name or MaterialClotrimazole
TypeClotrimazole
Physical FormPowder
CAS23593-75-1
Health Hazard 1H302-H315-H319
View more
Clotrimazole is an imidazole derivative and an antifungal compound and a CYP (cytochrome P450) inhibitor. Clotrimazole has been shown to block the intermediate-conductance, IK1 channels (Ca2+ activated K+ channels), in cells such as erythrocytes. In vitro studies of various yeast strains have demonstrated susceptibility to clotrimazole. Clotrimazole is an activator of MB67 and an inhibitor of CYP3A4 and CYP51A1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Clotrimazole is an imidazole derivative and an antifungal compound and a CYP (cytochrome P450) inhibitor. Clotrimazole has been shown to block the intermediate-conductance, IK1 channels (Ca2+ activated K+ channels), in cells such as erythrocytes. In vitro studies of various yeast strains have demonstrated susceptibility to clotrimazole. Clotrimazole is an activator of MB67 and an inhibitor of CYP3A4 and CYP51A1.

Solubility
Soluble in chloroform, DMSO, DMF and alcohols

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. E.B Souto.; b, S.A Wissing.; C.M Barbosa.; R.H Müller. Development of a controlled release formulation based on SLN and NLC for topical clotrimazole delivery.Int. J. Pharm. 2004, 278 (1),71-77 .
  2. Jack D. Sobel.; Doris Brooker.; Gary E. Stein.; Jessica L. Thomason.; Daniel P. Wermeling.; Blake Bradley.; Louis Weinstein. Single oral dose fluconazole compared with conventional clotrimazole topical therapy of Candida vaginitis.American Journal of Obstetrics and Gynecology. 1995, 172 (4),1263-1268 .