Thermo Scientific Chemicals

Colcemid, 98+%

Catalog number: J63900.MF
50 mg, Each
Thermo Scientific Chemicals

Colcemid, 98+%

Catalog number: J63900.MF
50 mg, Each
Quantity
Catalog number: J63900.MF
also known as J63900-MF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
477-30-5
IUPAC Name
3,4,5,14-tetramethoxy-10-(methylamino)tricyclo[9.5.0.0²,⁷]hexadeca-1(16),2,4,6,11,14-hexaen-13-one
Molecular Formula
C21H25NO5
InChI Key
NNJPGOLRFBJNIW-UHFFFAOYNA-N
SMILES
CNC1CCC2=CC(OC)=C(OC)C(OC)=C2C2=CC=C(OC)C(=O)C=C12
Assay (HPLC)
≥98.0%
Proton NMR
Conforms
Appearance (Color)
Pale yellow
Form
Powder

Description

Microtubule modulator that depolymerizes microtubules and inactivates spindle fiber formation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text