Phorbol 12-myristate 13-acetate, >94%, Thermo Scientific Chemicals
Phorbol 12-myristate 13-acetate, >94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phorbol 12-myristate 13-acetate, >94%, Thermo Scientific Chemicals

Activates protein kinase C and promotes skin tumors in mice.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
5 mg
10 mg
25 mg
Catalog number J63916.MCR
also known as J63916-M^
Price (USD)/ Each
94.65
Online exclusive
105.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (USD)/ Each
94.65
Online exclusive
105.00 
Save 10.35 (10%)
Add to cart
Phorbol 12-myristate 13-acetate, >94%, Thermo Scientific Chemicals
Catalog numberJ63916.MCR
Price (USD)/ Each
94.65
Online exclusive
105.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS16561-29-8
IUPAC Name(1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate
Molecular FormulaC36H56O8
InChI KeyPHEDXBVPIONUQT-RGYGYFBISA-N
SMILESCCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay from Suppliers CofA≥94.0%
FormPowder, film or foam, solid
PMA activates Ca2+- ATPase and potentiates forskolin-induced cAMP formation. It has been shown to inhibit apoptosis induced by the Fas antigen, but PMA induces apoptosis in HL-60 promyelocytic leukemia cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
PMA activates Ca2+- ATPase and potentiates forskolin-induced cAMP formation. It has been shown to inhibit apoptosis induced by the Fas antigen, but PMA induces apoptosis in HL-60 promyelocytic leukemia cells.

Solubility
Very soluble in methanol, soluble in DMSO, chloroform, ethanol or acetone, ether, ethyl acetate, DMF or acetonitrile. Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Vicentini, L.M.; Di Virgilio, F.; Ambrosini, A.; Pozzan, T.; Meldolesi, J. Tumor promoter phorbol 12-myristate, 13-acetate inhibits phosphoinositide hydrolysis and cytosolic Ca2+ rise induced by the activation of muscarinic receptors in PC12 cells. Biochemical and Biophysical Research Communications. 1985, 127, (1), 310-317.
  2. Emerit, I.; Cerutti, P.A. Tumor promoter phorbol 12-myristate 13-acetate induces a clastogenic factor in human lymphocytes. Current Issue. 1982, 79, (23), 7509-7513.