The primary use of L-phenylalanine in the body is in the production of tyrosine, another amino acid. It is an essential amino acid, which is biologically converted into L-tyrosine, another one of the DNA-encoded amino acids, which in turn is converted to L-DOPA and further converted into dopamine, norepinephrine, and epinephrine. L-Phenylalanine is produced for medical, feed, and nutritional applications such as in the preparation of Aspartame.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The primary use of L-phenylalanine in the body is in the production of tyrosine, another amino acid. It is an essential amino acid, which is biologically converted into L-tyrosine, another one of the DNA-encoded amino acids, which in turn is converted to L-DOPA and further converted into dopamine, norepinephrine, and epinephrine. L-Phenylalanine is produced for medical, feed, and nutritional applications such as in the preparation of Aspartame.
Solubility
Soluble in water. Aqueous solubility increases with warming. Also soluble in methanol, ethanol or 1M NH{4}OH
Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only
Jason W. Chin.; Stephen W. Santoro.; Andrew B. Martin.; David S. King.; Lei Wang.; Peter G. Schultz. Addition of p-Azido-l-phenylalanine to the Genetic Code of Escherichia coli.J. Am. Chem. Soc. 2002, 124 (31),9026-9027 .
F. Ko.; Y. Gogotsi.; A. Ali.; N. Naguib.; H. Ye.; G.L. Yang.; C. Li.; P. Willis. Electrospinning of Continuous Carbon Nanotube-Filled Nanofiber Yarns. J. Adv. Mater. 2003, 15 (14),1161-1165 .